missing translation for 'onlineSavingsMsg'
Learn More

Iodoacetic Anhydride, 97%

Product Code. 10043720 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10043720 250 mg 250mg
15179263 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10043720 Supplier Thermo Scientific Acros Supplier No. 307592500
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 54907-61-8 | C4H4I2O3 | 353.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54907-61-8
Molecular Formula C4H4I2O3
Molecular Weight (g/mol) 353.88
MDL Number MFCD00001080
InChI Key RBNSZWOCWHGHMR-UHFFFAOYSA-N
Synonym iodoacetic anhydride, di iodoacetic anhydride, acmc-20akq5, iodoacetic acid anhydride, 2-iodoacetyl 2-iodoacetate, acetic acid, iodo-, anhydride, acetic acid, 2-iodo-,1,1'-anhydride, iodoacetic anhydride, technical t
PubChem CID 2733254
IUPAC Name (2-iodoacetyl) 2-iodoacetate
SMILES C(C(=O)OC(=O)CI)I

Specifications

Melting Point 43.0°C to 49.0°C
Color Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (ICH2CO)2O
Quantity 250 mg
Formula Weight 353.88
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Iodoacetic anhydride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.