missing translation for 'onlineSavingsMsg'
Learn More

Iodocyclopentane, 97%, stab. with copper

Product Code. 11391307
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11391307 5 g 5g
11301317 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11391307 Supplier Thermo Scientific Alfa Aesar Supplier No. L01522.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1556-18-9 | C5H9I | 196.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1556-18-9
Molecular Formula C5H9I
Molecular Weight (g/mol) 196.03
MDL Number MFCD00039446
InChI Key PCEBAZIVZVIQEO-UHFFFAOYSA-N
Synonym cyclopentane, iodo, cyclopentyl iodide, jodcyclopentan, cyclopentyliodide, iodo-cyclopentane, iodocyclo-pentane, iodocyclopentane, acmc-1c0qu
PubChem CID 73784
IUPAC Name iodocyclopentane
SMILES IC1CCCC1

Specifications

Density 1.695
Boiling Point 166°C to 167°C
Flash Point 59°C (138°F)
Refractive Index 1.548
Quantity 5 g
UN Number UN1993
Beilstein 1900558
Sensitivity Light sensitive
Formula Weight 196.03
Percent Purity 97%
Chemical Name or Material Iodocyclopentane, stabilized with copper
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.