missing translation for 'onlineSavingsMsg'
Learn More

Iodoethane, Extra Pure, SLR, Fisher Chemical

Product Code. 10365850 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Plastic Coated Glass bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10365850 100 mL Plastic Coated Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10365850 Supplier Fisher Chemical Supplier No. I/0700/PB05
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

Especificaciones

CAS 75-03-6
Melting Point -108°C
Color Yellow
Boiling Point 69°C
Packaging Plastic Coated Glass bottle
Molecular Formula C2H5I
MDL Number 1091
Quantity 100 mL
Synonym ethyl iodide, ethane, iodo, monoiodoethane, hydriodic ether, ethyliodide, jodethan czech, ethyljodid, jodethan, iodo-ethane, ethyljodid czech
InChI Key HVTICUPFWKNHNG-UHFFFAOYSA-N
SMILES CCI
IUPAC Name iodoethane
Molecular Weight (g/mol) 155.966
PubChem CID 6340
ChEBI CHEBI:42487
Formula Weight 155.97g/mol
Physical Form Liquid
Mostrar más Mostrar menos
compliance-icons compliance-icons
Nombre del producto
  • Iodoethane, stabilized
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Consejos de prudencia
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.