missing translation for 'onlineSavingsMsg'
Learn More

Iodoethanol, TRC

Product Code. 30134282
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134282 250 mg 250mg
30137187 1 g 1g
30010650 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134282 Supplier Toronto Research Chemicals Supplier No. TRCI705875250MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-76-0
Molecular Formula C2 H5 I O
Molecular Weight (g/mol) 171.965
Synonym 2-Iodoethanol, 2-Hydroxy Iodide, 2-Iodoethan-1-ol, Ethylene Iodohydrin, NSC 85227
IUPAC Name 2-iodoethanol
SMILES OCCI

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C2H5IO/c3-1-2-4/h4H,1-2H2
Formula Weight 171.939
Chemical Name or Material Iodoethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.