missing translation for 'onlineSavingsMsg'
Learn More

Iodoform, Pure, SLR, Fisher Chemical

Product Code. 10548040 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic powder jar
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10548040 100 g Plastic powder jar 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10548040 Supplier Fisher Chemical Supplier No. I/0750/48
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 75-47-8
Melting Point 119°C
Color Yellow
Packaging Plastic powder jar
Molecular Formula CHI3
MDL Number 1069
Quantity 100 g
Synonym triiodomethane, methane, triiodo, carbon triiodide, jodoform, trijodmethane, dezinfekt v, jodoform czech, trijodmethane czech, ccris 346, unii-kxi2j76489
InChI Key OKJPEAGHQZHRQV-UHFFFAOYSA-N
SMILES C(I)(I)I
IUPAC Name iodoform
Molecular Weight (g/mol) 393.732
PubChem CID 6374
ChEBI CHEBI:37758
Formula Weight 393.72g/mol
Physical Form Solid
Show More Show Less
compliance-icons
Product Identifier
  • Iodoform
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.