missing translation for 'onlineSavingsMsg'
Learn More

Iodomethane, TRC

Product Code. 30005433
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005433 5 g 5g
30005422 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30005433 Supplier Toronto Research Chemicals Supplier No. TRCI7082005G
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74-88-4
Molecular Formula C H3 I
Molecular Weight (g/mol) 141.94
Synonym Methane, iodo-, Iodomethane, Ioguard, Methyl iodide, Methyl iodide (CH3I), Monoiodomethane, NSC 9366
IUPAC Name iodomethane
SMILES CI

Specifications

Quantity 5 g
InChI Formula InChI=1S/CH3I/c1-2/h1H3
Formula Weight 141.9279
Chemical Name or Material Methyl iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.