missing translation for 'onlineSavingsMsg'
Learn More

(Iodomethyl)cyclopropane, 95%

Product Code. 15497497
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497497 250 mg 250mg
15487497 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15497497 Supplier Thermo Scientific Alfa Aesar Supplier No. H35309.MD
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 33574-02-6 | C4H7I | 182.004 g/mol

(Iodomethyl)cyclopropane is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Iodomethyl)cyclopropane is used as pharmaceutical intermediate.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 33574-02-6
Molecular Formula C4H7I
Molecular Weight (g/mol) 182.004
MDL Number MFCD06410734
InChI Key JHKJQWFHNIOUKY-UHFFFAOYSA-N
Synonym iodomethyl cyclopropane, cyclopropylmethyl iodide, cyclopropane, iodomethyl, ambkkkkk549, acmc-20aplj, 2-iodomethyl cyclopropane, iodomethyl cyclopropane gc
PubChem CID 2763048
IUPAC Name iodomethylcyclopropane
SMILES C1CC1CI

Spezifikation

Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 182
Percent Purity 97%
Chemical Name or Material (Iodomethyl)cyclopropane

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.