missing translation for 'onlineSavingsMsg'
Learn More

Iodotrimethylsilane, 95-97%, stabilized, AcroSeal™

Product Code. 10530964 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10530964 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10530964 Supplier Thermo Scientific Acros Supplier No. 426421000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 16029-98-4 | C3H9ISi | 200.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 16029-98-4
Color Colorless to Red
Density 1.4000g/mL
Boiling Point 106°C
Flash Point -31°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (Argentometry)
Packaging AcroSeal™ Glass Bottle
Molecular Formula C3H9ISi
Linear Formula (CH3)3SiI
Refractive Index 1.4700 to 1.48
MDL Number MFCD00001028
Quantity 100 mL
Specific Gravity 1.4
Synonym trimethylsilyl iodide, trimethyliodosilane, silane, iodotrimethyl, iodo trimethyl silane, trimethylsilyliodide, tms-iodide, iodotrimethysilane, iodotrimetylsilane, tms iodide, iodo trimethylsilane
Solubility Information Solubility in water: reacts
InChI Key CSRZQMIRAZTJOY-UHFFFAOYSA-N
SMILES C[Si](C)(C)I
IUPAC Name iodo(trimethyl)silane
Stabilizer copper
Molecular Weight (g/mol) 200.1
PubChem CID 85247
Formula Weight 200.1
Percent Purity 95-97%
Physical Form Liquid
Chemical Name or Material Iodotrimethylsilane, Stabilized
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iodotrimethylsilane, stabilized
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
  • MIXTURE LIST-Contain: Iodotrimethylsilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.