missing translation for 'onlineSavingsMsg'
Learn More

Iodotrimethylsilane, 95-97%, stabilized

Product Code. 10583551 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
250 mL
Packaging:
Glass bottle
Unit Size:
100mL
250mL
25mL
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10583551 5 mL Glass bottle 5mL
10733001 25 mL Glass bottle 25mL
10583741 100 mL Glass bottle 100mL
13468428 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10583551 Supplier Thermo Scientific Acros Supplier No. 187490050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 16029-98-4 | C3H9ISi | 200.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16029-98-4
Molecular Formula C3H9ISi
Molecular Weight (g/mol) 200.1
MDL Number MFCD00001028
InChI Key CSRZQMIRAZTJOY-UHFFFAOYSA-N
Synonym trimethylsilyl iodide, trimethyliodosilane, silane, iodotrimethyl, iodo trimethyl silane, trimethylsilyliodide, tms-iodide, iodotrimethysilane, iodotrimetylsilane, tms iodide, iodo trimethylsilane
PubChem CID 85247
IUPAC Name iodo(trimethyl)silane
SMILES C[Si](C)(C)I

Specifications

Color Colorless to Red
Density 1.4000g/mL
Boiling Point 106.0°C
Flash Point −31°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (Ex I) (Argentometry)
Packaging Glass bottle
Linear Formula (CH3)3SiI
Refractive Index 1.4700 to 1.4800
Quantity 5 mL
Fieser 08,261; 09,251; 10,216; 11,271; 12,259; 13,329; 16,188; 06,81
Specific Gravity 1.4
Solubility Information Solubility in water: reacts
Stabilizer copper
Formula Weight 200.1
Percent Purity 95 to 97%
Physical Form Liquid
Chemical Name or Material Iodotrimethylsilane, 95-97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iodotrimethylsilane, stabilized
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
  • MIXTURE LIST-Contain: odotrimethylsilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.