missing translation for 'onlineSavingsMsg'
Learn More

Iodotrimethylsilane, 95-97%, stabilized

Product Code. 10583741 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
25 mL
100 mL
250 mL
Packaging:
Glass bottle
Unit Size:
100mL
250mL
25mL
5mL
This item is not returnable. View return policy

Product Code. 10583741

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16029-98-4 | C3H9ISi | 200.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16029-98-4
Molecular Formula C3H9ISi
Molecular Weight (g/mol) 200.1
MDL Number MFCD00001028
InChI Key CSRZQMIRAZTJOY-UHFFFAOYSA-N
Synonym trimethylsilyl iodide, trimethyliodosilane, silane, iodotrimethyl, iodo trimethyl silane, trimethylsilyliodide, tms-iodide, iodotrimethysilane, iodotrimetylsilane, tms iodide, iodo trimethylsilane
PubChem CID 85247
IUPAC Name iodo(trimethyl)silane
SMILES C[Si](C)(C)I

Specifications

Color Colorless to Red
Density 1.4000g/mL
Boiling Point 106.0°C
Flash Point −31°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (Ex I) (Argentometry)
Packaging Glass bottle
Linear Formula (CH3)3SiI
Refractive Index 1.4700 to 1.4800
Quantity 100 mL
Fieser 08,261; 09,251; 10,216; 11,271; 12,259; 13,329; 16,188; 06,81
Specific Gravity 1.4
Solubility Information Solubility in water: reacts
Stabilizer copper
Formula Weight 200.1
Percent Purity 95 to 97%
Physical Form Liquid
Chemical Name or Material Iodotrimethylsilane, 95-97%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.