missing translation for 'onlineSavingsMsg'
Learn More

Iodotrimethylsilane, 97%, stab. with copper

Product Code. 11498870
Click to view available options
Quantity:
5 g
25 g
100 g
250 g
Unit Size:
100g
250g
25g
5g
This item is not returnable. View return policy

Product Code. 11498870

Brand: Thermo Scientific Alfa Aesar A12902.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16029-98-4 | C3H9ISi | 200.094 g/mol

Iodotrimethylsilane is used for the introduction of trimethylsilyl group in organic synthesis. It is also useful for gas chromatography analysis by converting alcohol into a silyl ether derivative, thereby making it more volatile than the original molecule.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodotrimethylsilane is used for the introduction of trimethylsilyl group in organic synthesis. It is also useful for gas chromatography analysis by converting alcohol into a silyl ether derivative, thereby making it more volatile than the original molecule.

Solubility
Miscible with water.

Notes
Moisture and light Sensitive. Incompatible withstrong acids, strong bases, strong oxidizing agents and reacts violently with water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16029-98-4
Molecular Formula C3H9ISi
Molecular Weight (g/mol) 200.094
MDL Number MFCD00001028
InChI Key CSRZQMIRAZTJOY-UHFFFAOYSA-N
Synonym trimethylsilyl iodide, trimethyliodosilane, silane, iodotrimethyl, iodo trimethyl silane, trimethylsilyliodide, tms-iodide, iodotrimethysilane, iodotrimetylsilane, tms iodide, iodo trimethylsilane
PubChem CID 85247
IUPAC Name iodo(trimethyl)silane
SMILES C[Si](C)(C)I

Specifications

Boiling Point 105°C to 106°C
Quantity 25 g
Linear Formula (CH3)3SiI
UN Number UN2924
Beilstein 1731136
Flash Point −2°C (28°F)
Solubility Information Miscible with water.
Formula Weight 200.09
Percent Purity 97%
Refractive Index 1.476
Sensitivity Light and moisture sensitive
Density 1.44
Chemical Name or Material Iodotrimethylsilane, stabilized with copper
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iodotrimethylsilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
  • MIXTURE LIST-Contains : Iodotrimethylsilane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.