missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Iohexol

Product Code. 17231262 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17231262 25 g 25g
17241262 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17231262 Supplier Thermo Scientific Chemicals Supplier No. 466650250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 66108-95-0 | C19H26I3N3O9, C19H26I3N3O9 | 821.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66108-95-0
Molecular Formula C19H26I3N3O9

Specifications

Quantity 25 g
Solubility Solubility aq. soln.: soluble.
Melting Point 180.0°C
Formula Weight 821.13
InChI Key NTHXOOBQLCIOLC-UHFFFAOYNA-N
SMILES CC(=O)N(CC(O)CO)C1=C(I)C(C(=O)NCC(O)CO)=C(I)C(C(=O)NCC(O)CO)=C1I
IUPAC Name N1,N3-bis(2,3-dihydroxypropyl)-5-[N-(2,3-dihydroxypropyl)acetamido]-2,4,6-triiodobenzene-1,3-dicarboxamide
Molecular Weight (g/mol) 821.14
Chemical Name or Material Iohexol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.