missing translation for 'onlineSavingsMsg'
Learn More

Ion Torrent™ Ion AmpliSeq™ Library Kit 2.0

Product Code. 17152999
Change view
Click to view available options
No. of Reactions:
384 Reactions
Unit Size:
384 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
17152999 384 Reactions 384 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17152999 Supplier Ion Torrent™ Supplier No. 4480442

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The Ion AmpliSeq™ Library Kit 2.0 is designed for rapid preparation of amplicon libraries using Ion AmpliSeq™ panels.

The Ion AmpliSeq™ Library Kit 2.0 is designed for rapid preparation of amplicon libraries using Ion AmpliSeq™ panels.

Scalable Multiplex PCR Reactions

The Ion AmpliSeq 2.0 technology enables scalable multiplex PCR reactions from 12- to 24,000-plex in a single well using just 10 ng of starting DNA. Ion AmpliSeq panels and primer pools allow highly multiplexed PCR amplification of thousands of genomic target regions, with superior coverage uniformity and specificity, without the need of a specialized microfluidics PCR platform. In addition, the primers contain proprietary modifications that enable removal of primer sequences during library preparation, for efficient target assessment during sequencing. Multiple primer pools can be used to create overlapping amplicons that enable complete coverage of large targets. Ion AmpliSeq™ Custom Primer Pools are designed via the Ion AmpliSeq™ Designer Tool, available at www.ampliseq.com.

Barcoded Library Preparation

The Ion AmpliSeq Library Kit 2.0 includes reagents for generating amplicons with Ion AmpliSeq primers and preparing libraries from the resulting amplicons. The kit enables the preparation of barcoded libraries using Ion Xpress™ Barcode Adapters 1-96 kits or IonCode™ Barcode Adapters 1-384 kits. Barcoded libraries can be combined and loaded onto a single Ion chip to minimize the sequencing run time and cost and allow for accurate sample-to-sample comparisons.

Easy and Flexible Protocol

The Ion AmpliSeq Library Kit 2.0 uses a plate-based format for easier sample handling and tracking, and for compatibility with automation and high-throughput laboratories. The library preparation uses very small quantities of starting material per PCR. The resulting DNA libraries are ready for downstream template preparation using the automated Ion Chef™ System (which also provides the option of automated library preparation using the Ion AmpliSeq™ Kit for Chef DL8 (Cat. No. A29024)) or the Ion OneTouch™ 2 System followed by sequencing. The intuitive Torrent Suite™ Software and optional Ion Reporter™ Software enables automation of data analysis.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 7 tubes, store at -10°C to -30°C
1 tube Low TE Buffer, store at room temperature
Format Kit
Technique Library Generation
For Use With (Equipment) Ion GeneStudio™ S5 Series Sequencer, Ion Chef™ System
High-throughput Compatibility High-throughput Compatible, Multiplexing
Libraries Targeted Sequencing Library
Multiplex Capability 12- to 3072-plex per well
Product Line Ion AmpliSeq
Product Type Library Kit 2.0
Quantity 384 reactions
Sequencing Type Genome and DNA Sequencing
Species Human
For Use With (Application) Next-Generation Sequencing (NGS)
No. of Reactions 384 Reactions
Sample Type DNA (Genomic), DNA from FFPE Tissues, Tissue (FFPE)
Starting Material 10 ng gDNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.