Learn More
Stuart™ Ion Exchange Cartridges
Product Code. 16180921
Change view
Click to view available options
Capacity (Metric):
55 L
Unit Size:
Each
Product Code. Capacity (Metric) Unit Size
16180921 55 L Each
1 options
Product Code. 16180921 Supplier Stuart™ Supplier No. 8024493
In Stock
Add to basket
Add to basket
Stuart™ Ion Exchange Cartridges for use with Aquatron™ Water Stills
Aquatron™ Water Stills utilize disposable ion exchange cartridges to treat water
  • Cartridges slowly change colour from green to blue as they are exhausted to indicate the condition of the resin
  • Specifications
    For Use With (Equipment) Aquatron Water Stills
    Type Cartridge, Ion Exchange
    Capacity (Metric) 55 L