Learn More
Fisherbrand™ Ion Selective Electrode Iodide
Product Code. 11709718 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
pH Range:
0 to 14
Unit Size:
Each
Product Code. pH Range Unit Size
11709718 0 to 14 Each
1 options
Product Code. 11709718
  • ONLINE EXCLUSIVE PRICE
    488.00 €
    308.21 € / Pack of 1
    Save 179.79 €
    36% Off
View all promotions
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Combined ion-sensitive iodide electrode with solid membrane interchangeable flat surface is ideally delivered with membrane, electrolytic solution and gasket.
  • Combined ion-sensitive electrode with solid membrane interchangeable flat surface
  • The ion to be detected passes through the membrane generating a potential difference proportional to the ionic concentration
  • Analysis time typically less than one minute
  • Sample preparation steps such as filtering or distillation rarely necessary
  • Calibration should be carried out as often as possible
  • (less once a day) at the same temperature as the sample
  • Measuring temperature 0 to 50 ° C
Specifications
Description Ion Selective Electrode Iodide
pH Range 0 to 14