Learn More
Cole-Parmer™ Combination Ion-Selective Electrode (ISE)
Product Code. 15220748 Shop All Cole-Parmer Products
Change view
Click to view available options
Material:
Epoxy
Unit Size:
Each
Product Code. Material Unit Size
15220748 Epoxy Each
1 options
Product Code. 15220748 Supplier Cole-Parmer™ Supplier No. 2750434
  • ONLINE EXCLUSIVE PRICE
    364.00 €
    334.87 € / Pack of 1
    Save 29.13 €
    8% Off
Estimated Shipment: 05-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Fast and accurate Ion-selection electrodes do not require an external reference—they're easier to use than separate sensing and reference half-cell electrodes.
  • Cost-effective choices that are easy to use
  • Easily fit into small spaces—no additional reference electrode is needed
  • Water Hardness (Ca+²/Mg+²) ISE with epoxy body
  • Solid-state electrodes also have twelve polishing strips.
  • Electrodes are refillable
  • All electrodes include a 30-mL bottle of reference solution, a reference filling pipette, 1-m cable, BNC connector, and an instruction manual
Specifications
Description Combination Ion Selective Electrodes, Water hardness (Ca 2/Mg )
Material Epoxy