missing translation for 'onlineSavingsMsg'
Learn More

Ionomycin Calcium, TRC

Product Code. 30007465
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007465 25 mg 25mg
30003843 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30007465 Supplier Toronto Research Chemicals Supplier No. TRCI73124025MG
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56092-82-1
Molecular Formula C41CaH70O9
Molecular Weight (g/mol) 747.07
Synonym Calcium Ionomycin, (4R,6S,8S,10Z,12R,14R,16E,18R,19R,20S,21S)-11,19,21-Trihydroxy-4,6,8,12,14,18,20-heptamethyl-22-[(2S,2'R,5S,5'S)-octahydro-5'-[(1R)-1-hydroxyethyl]-2,5'-dimethyl[2,2'-bifuran]-5-yl]-9-oxo-10,16-Docosadienoic Acid Calcium Salt;
SMILES O[C@H](C)[C@]1(C)CC[C@]([H])([C@]2(O[C@H](C[C@H](O)[C@H](C)[C@H](O)[C@H](C)/C=C/C[C@H](C[C@H](/C(O[Ca+2]OC3=O)=C/C([C@@H](C)C[C@@H](C)C[C@H](C)CC3)=O)C)C)CC2)C)O1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C41H72O9.Ca/c1-25(21-29(5)34(43)24-35(44)30(6)22-27(3)20-26(2)14-15-38(46)47)12-11-13-28(4)39(48)31(7)36(45)23-33-16-18-41(10,49-33)37-17-19-40(9,50-37)32(8)42;/h11,13,24-33,36-37,39,42-43,45,48H,12,14-23H2,1-10H3,(H,46,47);/q;+2/p-2/b13-11+,34-24-;/t25-,26-,27+,28-,29-,30+,31+,32-,33+,36+,37-,39-,40+,41+;/m1./s1
Chemical Name or Material Ionomycin Calcium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.