missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Ionomycin free acid

Product Code. 18034772
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18034772 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18034772 Supplier Tocris Bioscience™ Supplier No. 2092/1
  • ONLINE EXCLUSIVE PRICE
    266.00 €
    250.95 € / 1mg
    Save 15.05 €
    5% Off
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Calcium ionophore

Ionomycin free acid is a calcium ionophore; more specific than A23187 .

Calcium Salt also available.

TRUSTED_SUSTAINABILITY

Specifications

CAS 56092-81-0
Quantity 1 mg
Molecular Formula C41H72O9
Formula Weight 709.01
Product Type Ionomycin Free Acid
Purity >98%
Molecular Weight (g/mol) 709.02
Chemical Name or Material Ionomycin free acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.