missing translation for 'onlineSavingsMsg'
Learn More

IOX1, MedChemExpress

Product Code. 16491317
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491317 50 mg 50mg
16425088 10 mg 10mg
16417807 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16491317 Supplier Medchem Express Supplier No. HY12304
 Restricted Item
Estimated Shipment: 30-11-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

MedChemExpress IOX1, 5-Carboxy-8-hydroxyquinoline, is a potent broad‐spectrum inhibitor of 2OG oxygenases, including the JmjC demethylases. IOX1 inhibits KDM4C, KDM4E, KDM2A, KDM3A and KDM6B with IC50 values of 0.6 μM, 2.3 μM, 1.8 μM, 0.1 μM and 1.4 μM, respectively. IOX1 also inhibits ALKBH5.

IOX1, 5-Carboxy-8-hydroxyquinoline, is a potent broad‐spectrum inhibitor of 2OG oxygenases, including the JmjC demethylases. IOX1 inhibits KDM4C, KDM4E, KDM2A, KDM3A and KDM6B with IC50 values of 0.6 μM, 2.3 μM, 1.8 μM, 0.1 μM and 1.4 μM, respectively. IOX1 also inhibits ALKBH5.

Specifica

Chemical Name or Material IOX1
CAS 5852-78-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C10H7NO3
Quantity 50 mg
Solubility Information DMSO : 13.33 mg/mL (70.47 mM; Need ultrasonic and warming)
SMILES O=C(C1=C2C=CC=NC2=C(O)C=C1)O
Molecular Weight (g/mol) 189.17
Formula Weight 189.17
Percent Purity 98.38%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Vedi altri risultati Mostra meno risultati

Research purposes only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.