missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ IP7e

Description
IP7e is a potent Nurr1 activator (EC50 = 3.9 nM). Delays onset and reduces severity of symptoms in mice with experimental autoimmune encephalomyelitis (EAE). Also suppresses NF-κB signaling. Brain penetrant and orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 6-[4-[(2-Methoxyethoxy)methyl]phenyl]-5-methyl-3-phenyl-isoxazolo[4,5-c]pyridin-4(5H)-one |
| CAS | 500164-74-9 |
| Quantity | 50 mg |
| Target | NFkB/IkB Inhibitors |
| Molecular Formula | C23H22N2O4 |
| Formula Weight | 390.43 |
| Purity | >98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?