missing translation for 'onlineSavingsMsg'
Learn More

AGMA IPA 70% in WFI Disinfectant Economy Wipe

Product Code. 30066740
Change view
Click to view available options
Dimensions (L x W):
9.84 x 9.84 in.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
30066740 9.84 x 9.84 in. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30066740 Supplier AGMA Supplier No. 1213
  • ONLINE EXCLUSIVE PRICE
    102.00 €
    99.43 € / Pack of 10
    Save 2.57 €
    2% Off
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Keep your Cleanroom environment clean and protected with our 0.2μm filtered 70% IPA in WFI disinfectant sprays and wipes.

The 0.2 μm filtered IPA 70% in WFI quality water is perfect as a proven industry standard biocide within pharmaceutical and hospital cleanrooms.

Trigger sprays are commonly used in cleanrooms, they are a high cost-effective means of routine disinfection when used with Dry Wipes. Designed for ease of use by operators who prefer a compact size of Trigger Spray.

All products are fully traceable with accompanying certification and with a 2 year unopened shelf life.

Perfect for day-to-day disinfection across a range of surfaces including worktops, instruments, gloved hands and transferred items.

  • Efficacy independently validated
  • 0.2 μm filtered and blended with WFI quality water - high purity water
  • Ergonomic trigger with effective return mechanism
  • Manufactured in Grade D cleanroom
  • Full traceability with supporting analysis
  • 2 - year shelf life
  • Also available in 500 mL and 1 L trigger sprays, 5 L and wet wipe formats
  • EN14476, EN1276, EN1650 and EN13697

Specifications

Product Type Disinfectant Wipe
Length (English) 9.84 in.
Length (Metric) 25 cm
Width (English) 9.84 in.
Width (Metric) 25 cm
Material Polypropylene
Autoclavable Not Autoclavable
Container Type Pouch
Disposable Limited-use
Dimensions (L x W) 9.84 x 9.84 in.
For Use With (Application) Cleanrooms and/or controlled environments subject to your contamination control strategies
Packaging Quantity 10 x 50 Wipes
Saturation Solution Isopropanol 70% v/v in WFI
Sterility Non-sterile
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • IPA 70% in WFI
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: propan-2-ol

Use biocides safely. Always read the label and product information before use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.