missing translation for 'onlineSavingsMsg'
Learn More

Chemsplash IPA Surface Isopropanol Disinfectant Wipe

Product Code. 30058236
Change view
Click to view available options
Dimensions (L x W):
20 x 20 cm
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
30058236 20 x 20 cm Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30058236 Supplier Chemsplash Supplier No. 4900
Stock Up and Save Up to 60%
Use promo code 28009
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

70% isopropanol Disinfectant Surface wipe, supplied in a self dispensing tub of 200 sheets. Each wipes is 20x20 cm.

70% Isopropanol Wet Wipes

  • 22GSM Strong Polypropylene Fabric
  • Efficient Against Surface Disinfection
  • Antibacterial, Antiviral and Effective Against Coronavirus
  • 200 Wipes Per Tub
  • Wipe Size: 20 x20 cm

Description: These 70% isopropanol disinfectant surface wipes come in a self-dispensing tub of 200 sheets. Each wipe measures 20x20cm and is designed for effective surface disinfection. The wipes are efficient against bacteria, viruses, and particularly effective against Coronavirus, offering a quick and convenient cleaning solution.

Specifications

Product Type Antibacterial Wipe
Length (English) 20 cm
Length (Metric) 20 cm
Width (English) 20 cm
Width (Metric) 20 cm
Material Polypropylene
Autoclavable Not Autoclavable
Disposable Single-use
Dimensions (L x W) 20 x 20 cm
For Use With (Application) Surface Wiping
Ply 1-ply
Sterility Sterile
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • X12 Chemsplash IPA Surface Wipe White One Size Tub of 200
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Isopropanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.