missing translation for 'onlineSavingsMsg'
Learn More

Ipamorelin, TRC

Product Code. 30404739
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30404739 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30404739 Supplier Toronto Research Chemicals Supplier No. TRCI7394202.5MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ipamorelin
CAS 170851-70-4
Molecular Formula C38 H49 N9 O5
Quantity 2.5 mg
Synonym Ipamorelin, L-Lysinamide, 2-methylalanyl-L-histidyl-3-(2-naphthalenyl)-D-alanyl-D-phenylalanyl-, 2-Methylalanyl-L-histidyl-3-(2-naphthalenyl)-D-alanyl-D-phenylalanyl-L-lysinamide, 696: PN: WO2004005342 PAGE: 46 claimed protein, Ipamorelin, NNC 26-0161
InChI Formula InChI=1S/C38H49N9O5/c1-38(2,41)37(52)47-32(21-28-22-42-23-43-28)36(51)46-31(20-25-15-16-26-12-6-7-13-27(26)18-25)35(50)45-30(19-24-10-4-3-5-11-24)34(49)44-29(33(40)48)14-8-9-17-39/h3-7,10-13,15-16,18,22-23,29-32H,8-9,14,17,19-21,39,41H2,1-2H3,(H2,40,48)(H
SMILES CC(C)(N)C(=O)N[C@@H](Cc1c[nH]cn1)C(=O)N[C@H](Cc2ccc3ccccc3c2)C(=O)N[C@H](Cc4ccccc4)C(=O)N[C@@H](CCCCN)C(=O)N
IUPAC Name (2S)-6-amino-2-[[(2R)-2-[[(2R)-2-[[(2S)-2-[(2-amino-2-methylpropanoyl)amino]-3-(1H-imidazol-4-yl)propanoyl]amino]-3-naphthalen-2-ylpropanoyl]amino]-3-phenylpropanoyl]amino]hexanamide
Molecular Weight (g/mol) 711.853
Formula Weight 711.386
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.