Learn More
Cytiva IPG Buffers
Produce more uniform conductivity along the Immobiline™ DryStrip during focusing. Cytiva IPG Buffers are ampholyte-containing buffers formulated for use with Immobiline™ DryStrip gels.
Brand: Cytiva 17-6000-86
66.89 EUR valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Code : GE
Additional Details : Weight : 0.10000kg
Description
- Eliminates background staining
- Use with Immobiline DryStrip gels to improve protein solubility
- Ensures optimal conditions for IEF separation
- High sample loading capacity, improved resolution, and increases the probability of detecting low abundance proteins when used with Immobiline DryStrip gels
Specifications
Colorless to Light Yellow | |
4 to 7 | |
Liquid | |
1mL |
Immobiline DryStrip Gels | |
IPG Buffer | |
IEF, 2-D Electrophoresis | |
2° to 8°C |