missing translation for 'onlineSavingsMsg'
Learn More

Cytiva IPG Buffers

Product Code. 10554565 Shop All Cytiva Products
Change view
Click to view available options
pH Range:
3 to 10
3 to 10 nonlinear
3 to 11 nonlinear
4 to 7
6 to 11
Unit Size:
1mL
Each
Product Code. pH Range unitSize
10554565 6 to 11 1mL
10534565 4 to 7 1mL
10084334 3 to 10 1mL
10544565 3 to 10 nonlinear Each
10014104 3 to 11 nonlinear 1mL
5 options
This item is not returnable. View return policy

Product Code. 10554565

Brand: Cytiva 17600178

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Produce more uniform conductivity along the Immobiline™ DryStrip during focusing. Cytiva IPG Buffers are ampholyte-containing buffers formulated for use with Immobiline™ DryStrip gels.

  • Eliminates background staining
  • Use with Immobiline DryStrip gels to improve protein solubility
  • Ensures optimal conditions for IEF separation
  • High sample loading capacity, improved resolution, and increases the probability of detecting low abundance proteins when used with Immobiline DryStrip gels

Order Info

Supplied in 1mL aliquots of water; order Immobiline Drystrip gels separately.

TRUSTED_SUSTAINABILITY

Specifications

Color Colorless to Light Yellow
pH Range 6 to 11
Form Liquid
For Use With (Application) IEF, 2-D Electrophoresis
Quantity 1mL
Storage Requirements 2° to 8°C
Product Type IPG Buffer
For Use With (Equipment) Immobiline DryStrip Gels
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.