missing translation for 'onlineSavingsMsg'
Learn More

IPI 145, TRC

Product Code. 30407180
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30407180 100 mg 100mg
30405650 10 mg -
30404850 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30407180 Supplier Toronto Research Chemicals Supplier No. TRCI739400100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1201438-56-3
Molecular Formula C22 H17 Cl N6 O
Molecular Weight (g/mol) 416.86
Synonym 1(2H)-Isoquinolinone, 8-chloro-2-phenyl-3-[(1S)-1-(9H-purin-6-ylamino)ethyl]-, 8-Chloro-2-phenyl-3-[(1S)-1-(9H-purin-6-ylamino)ethyl]-1(2H)-isoquinolinone, Copiktra, Duvelisib, INK 1197, IPI 145
IUPAC Name 8-chloro-2-phenyl-3-[(1S)-1-(9H-purin-6-ylamino)ethyl]isoquinolin-1-one
SMILES C[C@H](Nc1ncnc2[nH]cnc12)C3=Cc4cccc(Cl)c4C(=O)N3c5ccccc5

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C22H17ClN6O/c1-13(28-21-19-20(25-11-24-19)26-12-27-21)17-10-14-6-5-9-16(23)18(14)22(30)29(17)15-7-3-2-4-8-15/h2-13H,1H3,(H2,24,25,26,27,28)/t13-/m0/s1
Formula Weight 416.1152
Chemical Name or Material IPI 145
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.