missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iPrep™ Card: gDNA Tissue

Product Code. 10409793
Change view
Click to view available options
Quantity:
1 card
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10409793 1 card Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10409793 Supplier Invitrogen™ Supplier No. IS10013
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

iPrep™ Card: gDNA Tissue

The iPrep™ Protocol Card is a flash, 512 KB memory card. Each iPrep™ Protocol card is pre-programmed with the purification protocol that directs the volume of reagents used and incubation time.

The following iPrep™ Protocol Cards are available from Invitrogen:

  • iPrep™ Forensic Card for use with the iPrep™ ChargeSwitch™ Forensic Kit and the iPRep™ ChageSwitch™ gDNA Buccal kit
  • iPrep™ Tissue Card for use with the iPrep ChargeSwitch™ Tissue Kit
  • iPrep™ Blood Card for use with the iPrep PureLink Blood kit

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
For Use With (Equipment) iPrep™
Product Line iPrep
Quantity 1 card
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.