missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iprodione-d5 (3,5-dichlorophenyl-2,4,6-d3; hydantoin-5,5-d2), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Iprodione-d5 |
| CAS | 1215631-57-4 |
| Molecular Formula | C13 2H5 H8 Cl2 N3 O3 |
| Quantity | 0.01 g |
| InChI Formula | InChI=1 S/C13H13Cl2N3O3/c1-7(2)16-12(20)17-6-11(19)18(13(17)21)10-4-8(14)3-9(15)5-10/h3-5,7 H,6H2,1-2H3,(H,16,20)/i3D,4 D,5 D,6D2 |
| SMILES | [2 H]c1c(Cl)c([2 H])c(N2C(=O)N(C(=O)NC(C)C)C([2 H])([2 H])C2=O)c([2 H])c1Cl |
| IUPAC Name | 5,5-dideuterio-3-(3,5-dichloro-2,4,6-trideuteriophenyl)-2,4-dioxo-N-propan-2-ylimidazolidine-1-carboxamide |
| Molecular Weight (g/mol) | 335.2 |
| Formula Weight | 334.0648 g/mol |
| Percent Purity | 97 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction