Learn More
Thermo Scientific™ TheraPure™ iPrPAC-rG Phosphoramidite, 20-400 finish bottle
Shop All Thermo Scientific ProductsDescription
iPrPAC-rG CEP is a phosphoramidite containing 5' DMT and the nucleoside guanosine (G) with Isopropylphenoxyacetyl (iPrPAC) base protection, for use in oligonucleotide synthesis. RNA phosphoramidites contain a 2'-OH protecting group, tBDMS (tert-butyl dimethylsilyl). Each base has unique protecting groups designed specifically to protect the RNA during the chemical synthesis process, and produce the highest amount of full length product. Alternate base protection options are also available to allow for faster deprotection and longer oligonucleotide synthesis. The TheraPure trademark denotes our highest level of phosphoramidite purity, developed with the greatest stringency of high definition analytics and tightly controlled impurity profiles and, therefore, ideal for any therapeutic application. This product is supplied in a 30 mL Amber bottle with a 20-400 Finish neck style, which is compatible with most Expedite Systems of oligonucleotide synthesizers.
Features:
• Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-N2-isopropylphenoxyacetyl-2'-O-TBDMSi-guanosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula: C57H74N7O10PSi
• Base: rG
• Base Protection: iPrPAC
• Formula weight: 1076.3 g/mol
• CAS number: 956580-73-7
• HPLC Purity: ≥99%
• 31P NMR Purity: ≥99%; Sum of non-primary peaks in 140-152 ppm range: ≤0.5%; No single PIII impurity (140-152 ppm) >0.3%
Specifications
Specifications
| Format | Powder |
| For Use With (Equipment) | Expedite Synthesizers |
| Product Type | iPrPAC-rG CEP |
| Content And Storage | Store in freezer (at or below -20°C). |
| Nucleoside Type | Guanosine (G) |
| Reagent Features | Fast deprotection |
| Shipping Condition | Room Temperature |
| Purity | 99% |
| Quantity | 0.5 g |
| Reagent Type | Phosphoramidite |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.