missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford IPTG
Description
IPTG is combined with X-GAL to detect lac+ colonies or cells in a colorimetric assay.
IPTG is combined with X-GAL to detect lac+ colonies or cells in a colorimetric assay. Distinguishes recombinants (white) from non- recombinants (blue) in cloning strategies using vectors such as Lambda-11, M13mp 18 and 19, pUC18 and 19, pUR222 containing the lacZ gene.
Dioxane-Free
Water Content: <0.49%Specifications
Specifications
| CAS | 367-93-1 |
| Molecular Formula | C9H18O5S |
| Quantity | 5 g |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?