missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ IPTG

Product Code. 18771468
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18771468 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18771468 Supplier R&D Systems™ Supplier No. 5509/10G
  • ONLINE EXCLUSIVE PRICE
    572.94 €
    514.50 € / 10g
    Save 58.44 €
    10% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Used in cloning procedures with X-GAL

IPTG is an used in combination with X-GAL (Cat. No. 5510) in cloning procedures requiring expression of β-galactosidase activity. Non-metabolizable analog of galactose.

This product has <0.1% dioxane content.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Methylethyl 1-thio-β-D-galactopyranoside
CAS 367-93-1
Synonym 1-Methylethyl 1-thio-β-D-galactopyranoside
Quantity 10 g
Solubility Soluble to 100mM in water
Product Line Tocris
Target Buffers, Chelators and Reagents
Molecular Formula C9H18O5S
Formula Weight 238.3
Physical Form Solid
Product Type Bioactive Small Molecule
Purity 99%
Show More Show Less

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.