missing translation for 'onlineSavingsMsg'
Learn More

Melford IPTG [Isopropyl-β-D-thiogalactopyranoside] Non mammalian

Product Code. 30017925
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30017925 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30017925 Supplier Melford Supplier No. 2946I56000100.0
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

IPTG is combined with X-GAL to detect lac+ colonies or cells in a colorimetric assay. Distinguishes recombinants (white) from non- recombinants (blue) in cloning strategies using vectors such as Lambda-11, M13mp 18 and 19, pUC18 and 19, pUR222 containing the lacZ gene.

Specifications

Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.