missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies IQ-1
Description
- IQ-1 selectively inhibits p300-dependent β-catenin signaling by binding to the PR72/130 subunit of protein phosphatase PP2A, leading to decreased phosphorylation of the β-catenin coactivator, p300, and decreased affinity of p300 for β-catenin
- IQ-1 thereby inhibits β-catenin/p300 interaction while increasing β-catenin/CBP mediated transcription (Miyabayashi et al.)
- Used with Wnt3a to maintain pluripotency of mouse embryonic stem (ES) cells in the absence of mouse embryonic fibroblasts (MEFs), serum, or exogenous leukemia inhibitory factor (LIF; Miyabayashi et al.)
- Enhances expansion of mouse ES-derived cardiovascular progenitor cells (Schenke-Layland et al.)
- Induces the conversion of cancer cells to a side population of cancer stem-like cells with high levels of drug resistance and tumorigenicity (He et al.)
Specifications
Specifications
| CAS | 331001-62-8 |
| Quantity | 5 mg |
| Molecular Formula | C21H22N4O2 |
| Purity | ≥98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?