Learn More
Memmert™ Iq Document With Device-Specific Works Test Data For Validation By Customer, For Hcp105
Includes IQ document with works test data for chamber.
Brand:  Memmert™ HCP105D00124
Additional Details : Weight : 0.01100kg
Product Code. 16394787
  • 452.00 € / Each
Estimated Shipment: 18-06-2024
to see stock.
Includes: Works test data
Description
Description
  • OQ/PQ check list as support for validation by customer
Specifications
Specifications
30, 55, 75, 110, 160, 260, 450, 750, 1060 Models Climate Chamber
IQ Document