Learn More
Tocris Bioscience™ Irbesartan
GREENER_CHOICE
Potent AT1 antagonist
Brand:  Tocris Bioscience™ 5798/50
Encompass
Product Code. 15797116
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Irbesartan is a potent angiotensin II type 1 receptor (AT1) antagonist (IC50 = 1.3 nM). Displays antihypertensive activity. Orally bioavailable.
Specifications
Specifications
2-Butyl-3-[[2′-(2H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-1,3-diazaspiro[4.4]non-1-en-4-one
138402-11-6