missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Irbesartan

Product Code. 18715318
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18715318 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18715318 Supplier Tocris Bioscience™ Supplier No. 5798/50
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent AT1 antagonist

Irbesartan is a potent angiotensin II type 1 receptor (AT1) antagonist (IC50 = 1.3 nM). Displays antihypertensive activity. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Butyl-3-[[2′-(2H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-1,3-diazaspiro[4.4]non-1-en-4-one
CAS 138402-11-6
Quantity 50 mg
Target Angiotensin AT1 Receptor Antagonists
Molecular Formula C25H28N6O
Formula Weight 428.53
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.