missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Iressa

Product Code. 30001338
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001338 10 mg 10mg
18093942 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 30001338 Supplier Tocris Bioscience™ Supplier No. 3000/10
  • ONLINE EXCLUSIVE PRICE
    246.00 €
    232.05 € / 10mg
    Save 13.95 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

Orally active, selective EGFR inhibitor

Iressa is an orally active, selective inhibitor of EGFR tyrosine kinase (IC50 = 23 - 79 nM). Shows minimal activity against ErbB2, KDR, c-flt, PKC, MEK and ERK-2. Blocks EGFR autophosphorylation and inhibits tumor growth in mice bearing a range of human xenografts. Also inhibits growth of brain metastases in mouse model of non-small cell lung cancer. View information regarding the usage of Iressa for non-clinical studies .
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 184475-35-2
Molecular Formula C22H24ClFN4O3

Specifications

Quantity 10 mg
Formula Weight 446.9
Purity >98%
Molecular Weight (g/mol) 446.91
Chemical Name or Material N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-[3-(4-morpholinyl)propoxy]-4-quinazolinamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.