missing translation for 'onlineSavingsMsg'
Learn More

Iridium, 99.9%, (trace metal basis), powder

Product Code. 10614712 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
10 g
Packaging:
Glass bottle
Unit Size:
10g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10614712 500 mg Glass bottle 500mg
10713534 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10614712 Supplier Thermo Scientific Acros Supplier No. 198095000
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 7439-88-5 | Ir | 192.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-88-5
Molecular Formula Ir
Molecular Weight (g/mol) 192.22
MDL Number MFCD00011062
InChI Key GKOZUEZYRPOHIO-UHFFFAOYSA-N
Synonym ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge
PubChem CID 23924
ChEBI CHEBI:49666
IUPAC Name iridium
SMILES [Ir]

Specifications

Color Gray
Quantity 500 mg
Packaging Glass bottle
Assay Percent Range 99.9%
Form Amorphous Powder
Fieser 02,228; 09,257
Merck Index 15, 5129
Formula Weight 192.2
Grade Trace Metal Basis
Chemical Name or Material Iridium
compliance-icons compliance-icons
Product Identifier
  • Iridium, powder
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 1
Hazard Statement
  • H319-Causes serious eye irritation.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.