missing translation for 'onlineSavingsMsg'
Learn More

Irinotecan hydrochloride trihydrate, 99%, Thermo Scientific™

Topoisomerase I inhibitor

Brand:  Thermo Scientific Alfa Aesar J62370.MC


Quantity 100mg

 View more versions of this product

Product Code. 15424769

  • 174.00 € / 100mg
 Restricted Item
Estimated Shipment: 29-09-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

136572-09-3
623.15
GURKHSYORGJETM-WAQYZQTGSA-N
74990
hydrogen (19S)-10,19-diethyl-19-hydroxy-14,18-dioxo-17-oxa-3,13-diazapentacyclo[11.8.0.0²,¹¹.0⁴,⁹.0¹⁵,²⁰]henicosa-1(21),2(11),3,5,7,9,15(20)-heptaen-7-yl [1,4'-bipiperidine]-1'-carboxylate chloride
C33H39ClN4O6
MFCD01862255
irinotecan hydrochloride, irinotecan hcl, topotecin, campto, cpt-11, cpt 11, camptothecin 11 hydrochloride, camptothecin 11, unii-06x131e4oe
CHEBI:5971
[H+].[Cl-].CCC1=C2C=C(OC(=O)N3CCC(CC3)N3CCCCC3)C=CC2=NC2=C1CN1C2=CC2=C(COC(=O)[C@]2(O)CC)C1=O

Specifications

136572-09-3
MFCD01862255
14,5091
Soluble in DMSO at 100mg/ml; Soluble in water at 25mg/ml with warming
[H+].[Cl-].CCC1=C2C=C(OC(=O)N3CCC(CC3)N3CCCCC3)C=CC2=NC2=C1CN1C2=CC2=C(COC(=O)[C@]2(O)CC)C1=O
623.15
CHEBI:5971
99%
C33H39ClN4O6
100mg
irinotecan hydrochloride, irinotecan hcl, topotecin, campto, cpt-11, cpt 11, camptothecin 11 hydrochloride, camptothecin 11, unii-06x131e4oe
GURKHSYORGJETM-WAQYZQTGSA-N
hydrogen (19S)-10,19-diethyl-19-hydroxy-14,18-dioxo-17-oxa-3,13-diazapentacyclo[11.8.0.0²,¹¹.0⁴,⁹.0¹⁵,²⁰]henicosa-1(21),2(11),3,5,7,9,15(20)-heptaen-7-yl [1,4'-bipiperidine]-1'-carboxylate chloride
74990
677.19 (623.15 Anhydrous)
Irinotecan hydrochloride trihydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Irinotecan hydrochloride trihydrate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H372-Causes damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contain: Irinotecan hydrochloride trihydrate

SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.