missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ IRIS11 Prestained Protein Ladder

Product Code. 30163837
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30163837 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30163837 Supplier Novus Biologicals™ Supplier No. NBP333168
  • ONLINE EXCLUSIVE PRICE
    149.00 €
    140.70 € / 500µL
    Save 8.30 €
    5% Off
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Prestained Protein Ladder

The IRIS11 Prestained Protein Ladder is a combination of 11 pre-stained proteins with molecular weights from 3 to 260 kDa. The 11 recombinant proteins are covalently coupled with blue chromophore, while 2 red bands at 70 kDa and 260 kDa, a green band at 15 kDa and a newly designed peacock green band at 60 kDa serve as reference bands. The IRIS11 Prestained Protein Ladder keeps track of the size and separation of proteins during SDS-polyacrylamide gel electrophoresis, approximating the target protein size and validating the Western transfer efficiency on PVDF, nylon, or nitrocellulose membranes.

TRUSTED_SUSTAINABILITY

Specifications

Formulation 20 mM Trisphosphate at pH 7.5, 2% SDS, 0.2 mM Dithiothreitol, 3.6 M Urea, and 15% (v/v) Glycerol.
For Use With (Application) 3ul or 5ul per loading for clear visualization during electrophoresis on 15-well or 10-well mini-gel, respectively. 2.5ul per well for general Western transferring.
Quantity 500 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.