missing translation for 'onlineSavingsMsg'
Learn More

Iron, 99%, powder, -70 mesh (<212 micron)

Product Code. 10771471 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
10 kg
Packaging:
Plastic bottle
Unit Size:
100g
10kg
2kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10771471 2 kg Plastic bottle 2kg
10217073 100 g Plastic bottle 100g
10193970 500 g Plastic bottle 500g
10454025 10 kg Plastic bottle 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10771471 Supplier Thermo Scientific Acros Supplier No. 197810020
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7439-89-6 | Fe | 55.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-89-6
Molecular Formula Fe
Molecular Weight (g/mol) 55.85
MDL Number MFCD00010999
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
Synonym ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
PubChem CID 23925
ChEBI CHEBI:82664
IUPAC Name iron
SMILES [Fe]

Specifications

Color Gray
Melting Point 1535.0°C
Boiling Point 3000.0°C
Density 7.8600g/mL
Quantity 2 kg
Packaging Plastic bottle
Assay Percent Range 99%
Form Fine Powder
Fieser 01,519; 02,229; 03,167; 05,357; 09,257; 14,184; 16,189; 17,157
Merck Index 15, 5137
Formula Weight 55.84
Percent Purity 99%
Chemical Name or Material Iron
Show More Show Less
compliance-icons
Product Identifier
  • Iron, powder
Signal Word
  • Danger
Hazard Category
  • Self-heating substance Category 1
Hazard Statement
  • H251-Self-heating: may catch fire.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P235+P410-Keep cool. Protect from sunlight.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.