missing translation for 'onlineSavingsMsg'
Learn More

Iron boride, 98% (metals basis)

Product Code. 11348227
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11348227

Brand: Thermo Scientific Alfa Aesar 022952.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12006-85-8 | BFe2 | 122.5 g/mol

Iron boride is used in the production of borated steels and borated stainless steels, deoxidizing agents, increase the hardness of steel, and alloying components of coatings applied to weld-facing electrodes. It is also used to improve abrasion resistance, wear resistance, and oxidation resistance. It is used in oil and gas refinery, chemical extraction, automotive, agricultural, stamping, textile extrusion and injection molding industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12006-85-8
Molecular Formula BFe2
Molecular Weight (g/mol) 122.5
MDL Number MFCD00016092
InChI Key FSDZRQFSRALZQJ-UHFFFAOYSA-N
Synonym boron; iron, diiron boride, iron boride fe2b, boranylidyne diiron
PubChem CID 6336853
IUPAC Name boron;iron
SMILES [B].[Fe].[Fe]

Specifications

Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 122.5
Percent Purity 98%
Odor Odorless
Chemical Name or Material Iron boride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.