missing translation for 'onlineSavingsMsg'
Learn More

Iron-chrome based high temperature gas shift catalyst, HiFUEL∣r W210

Product Code. 15476935
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Unit Size:
1kg
2.5kg
500g
This item is not returnable. View return policy

Product Code. 15476935

Brand: Thermo Scientific Alfa Aesar 045467.A4

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Iron-chrome based high temperature gas shift catalyst is ideal for use in fuel cell and other distributed hydrogen production applications. It is also used for fuel processing applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color Red to Brown
Quantity 2.5 kg
Solubility Information Insoluble in water.
Physical Form Pellets
Chemical Name or Material Iron-chrome based high temperature gas shift catalyst HiFUEL™ W210

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.