missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) acetate, anhydrous, Fe 29.5% min

Product Code. 11317418
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11317418 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11317418 Supplier Thermo Scientific Alfa Aesar Supplier No. 031140.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3094-87-9 | C4H6FeO4 | 173.93 g/mol

Iron(II) acetate is used as a mordant by the dye industry, as a catalyst in organic oxidation reactions and in carbon nanotube synthesis. It is used as a precursor in the production of other iron compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3094-87-9
Molecular Formula C4H6FeO4
Molecular Weight (g/mol) 173.93
MDL Number MFCD00058909
InChI Key LNOZJRCUHSPCDZ-UHFFFAOYSA-L
Synonym iron ii acetate, ferrous acetate, iron diacetate, iron di acetate, iron 2+ acetate, acetic acid, iron 2+ salt, acetic acid, iron 2+ salt 2:1, unii-l80i7m6d3q, ferrousacetate
PubChem CID 18344
SMILES [Fe++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 190°C to 200°C (decomposition)
Color Brown
Physical Form Lump/Powder
Quantity 25 g
Solubility Information Soluble in water.
IUPAC Name iron(2+);diacetate
Formula Weight 173.94
Odor Characteristic
Sensitivity Air and moisture sensitive
Chemical Name or Material Iron(II) acetate, anhydrous
compliance-icons
Product Identifier
  • Iron(II) acetate, anhydrous
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.