missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) bromide hydrate

Product Code. 11307418
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11307418

Brand: Thermo Scientific Alfa Aesar 031133.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13463-12-2 | Br2Fe | 215.65 g/mol

It is widely as a pharmaceutical intermediate.It is a useful synthetic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13463-12-2
Molecular Formula Br2Fe
Molecular Weight (g/mol) 215.65
MDL Number MFCD00016081
InChI Key GYCHYNMREWYSKH-UHFFFAOYSA-L
Synonym iron dibromide, unii-ea3x054rbz, iron ii bromide, ea3x054rbz, iron 2+ dibromide, iron ion 2+ dibromide, iron ii bromide, anhydrous 5g, iron ii bromide, ultra dry metals basis, ferrous bromide, anhydrous
PubChem CID 82240
IUPAC Name λ²-iron(2+) dibromide
SMILES [Fe++].[Br-].[Br-]

Specifications

Melting Point 27°C
Color Green
Physical Form Lump
Quantity 25 g
Linear Formula FeBr2·xH2O
UN Number UN3260
Solubility Information It is soluble in water,soluble in other solvents such as THF,methanol,ethanol.
Formula Weight 215.67 (Anhydrous)
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Density 3.9 g/mL
Chemical Name or Material Iron(II) bromide hydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iron(II) bromide hydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.