missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) chloride, anhydrous, 99.5% (metals basis)

Product Code. 11337418
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
Unit Size:
250g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11337418 5 g 5g
11357418 50 g 50g
11347418 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11337418 Supplier Thermo Scientific Alfa Aesar Supplier No. 031141.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7758-94-3 | Cl2Fe | 126.75 g/mol

Iron(II) chloride is used in the synthesis of iron complexes like iron(II) hydroxide. It acts as a precursor to hydrated iron(III) oxides that are magnetic pigments. It serves as a reducing agent in organic synthesis. Also, used as a reducing flocculating agent in wastewater treatment, especially for wastes containing chromate. It is used as an intermediate, in pigments, processing aids and in solids separation agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-94-3
Molecular Formula Cl2Fe
Molecular Weight (g/mol) 126.75
MDL Number MFCD00011004
InChI Key NMCUIPGRVMDVDB-UHFFFAOYSA-L
IUPAC Name λ²-iron(2+) dichloride
SMILES [Cl-].[Cl-].[Fe++]

Specifications

Boiling Point 1023°C
Melting Point 674°C
pH 2.5
Physical Form Powder
Quantity 5 g
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Density 3.160
Linear Formula FeCl2
UN Number UN3260
Merck Index 14,4043
Solubility Information Freely soluble in water,alcohol,acetone;Soluble in water,alcohol and acetone; Slightly soluble in benzene; Insoluble in ether.
Formula Weight 126.75
Percent Purity 99.5%
Assay (metals basis)
Chemical Name or Material Iron(II) chloride, anhydrous
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iron(II) chloride, anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/physician/if you feel unwell.
  • P310-Immediately call a POISON CENTER/doctor/physician.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.