missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) Disulfide, Extra Pure, SLR, Powder, Fisher Chemical

Product Code. 10636262 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 kg
Packaging:
Glass Jar
Unit Size:
1kg
This item is not returnable. View return policy

Product Code. 10636262

Brand: Fisher Chemical I/1165/60

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 1171°C
CAS 12068-85-8
Color Gray
Physical Form Powder/Solid
Quantity 1 kg
Molecular Formula FeS2
MDL Number MFCD00064690
Synonym iron disulfide, iron sulfide, pyrite, bis sulfanylidene iron, disulfanylideneiron, iron disulfide iron pyrite, marcasite, naturally occuring mineral, grains, approximately 0.06-0.19 in, pyrite, naturally occurring mineral, grains, approximately 0.06-0.19in
InChI Key NFMAZVUSKIJEIH-UHFFFAOYSA-N
SMILES S=[Fe]=S
IUPAC Name disulfanylideneiron
Molecular Weight (g/mol) 119.97
PubChem CID 14788
Formula Weight 119.967g/mol
Packaging Glass Jar
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.