missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) ethylenediammonium sulfate tetrahydrate, 99%

Product Code. 11337308
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337308 100 g 100g
11347308 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11337308 Supplier Thermo Scientific Alfa Aesar Supplier No. 030547.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 113193-60-5

Iron(II) ethylenediammonium sulfate tetrahydrate is used in the preparation of gold nanoparticles by reacting with gold(III)-bis[ethylene-diamine]-chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113193-60-5
MDL Number MFCD00082474

Specifications

Color Green to White
Linear Formula FeC2H4(NH3)2(SO4)2·4H2O
Quantity 100 g
Solubility Information Soluble in water.
Formula Weight 382.14 (310.08 Anhydrous)
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Iron(II) ethylenediammonium sulfate tetrahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.