missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) methoxide, 98%

Product Code. 11368057
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11368057 2 g 2g
11358057 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368057 Supplier Thermo Scientific Alfa Aesar Supplier No. 022552.04
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7245-21-8 | C2H6FeO2 | 117.913 g/mol

Iron(II) methoxide, is used in Engineering & Environmental, Catalysis & Synthesis, and In organic syntheses, chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7245-21-8
Molecular Formula C2H6FeO2
Molecular Weight (g/mol) 117.913
MDL Number MFCD00061474
InChI Key VAPRHKOWFRYFTF-UHFFFAOYSA-N
Synonym iron ii methoxide, iron 2+ dimethoxide, methanol, iron 2+ salt, acmc-1bc40
PubChem CID 14598007
IUPAC Name iron(2+);methanolate
SMILES C[O-].C[O-].[Fe+2]

Specifications

Color Green
Odor Alcohol-like
Linear Formula Fe(OCH3)2
Quantity 2 g
UN Number UN3181
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 117.92
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Iron(II) methoxide
Show More Show Less
compliance-icons compliance-icons
Produktidentifikator
  • Iron(II) methoxide
Signalwort
  • Warning
Gefahrenkategorie
  • Flammable solid Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H228-Flammable solid.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.