missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) oxalate dihydrate, Puratronic™, 99.999% (metals basis)

Product Code. 15445965
Change view
Click to view available options
Quantity:
2.5 g
25 g
100 g
500 g
Unit Size:
100g
2.5g
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15445965 25 g 25g
11330999 2.5 g 2.5g
11320999 100 g 100g
11310999 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15445965 Supplier Thermo Scientific Alfa Aesar Supplier No. 010718.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6047-25-2 | C2H4FeO6 | 179.89 g/mol

It is used as photographic developer for silver bromide-gelatin plates. It imparts greenish-brown tint to optical glass (sunglasses, windshields, railroad car windows), for decorative glassware, pigment for plastics, paints and lacquers. It acts as reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6047-25-2
Molecular Formula C2H4FeO6
Molecular Weight (g/mol) 179.89
MDL Number MFCD00150040 MFCD00012475
InChI Key NPLZZSLZTJVZSX-UHFFFAOYSA-L
Synonym ferrous oxalate dihydrate, unii-z6x3ybu50d, z6x3ybu50d, iron ii oxalate hydrate, iron ii oxalate dihydrate, iron oxalate dihydrate, iron 2+ ; oxalate; dihydrate, iron, diaqua ethanedioato 2--kappao1,kappao2-, t-4, iron ii oxalate dihydrate 250g, iron ii oxalate dihydrate, puratronic™
PubChem CID 516788
SMILES O.O.[Fe++].[O-]C(=O)C([O-])=O

Specifications

Melting Point 190°C (decomposition)
Physical Form Powder
Quantity 25 g
Solubility Information Soluble in water. Insoluble in acetic acid.
IUPAC Name iron(2+);oxalate;dihydrate
Formula Weight 179.90 (143.87 Anhydrous)
Percent Purity 99.999%
Grade Puratronic
Sensitivity Hygroscopic
Density 2.28 g/mL
Assay (metals basis)
Chemical Name or Material Iron(II) oxalate dihydrate
Show More Show Less
compliance-icons
Product Identifier
  • Iron(II) oxalate dihydrate, Puratronic
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P363-Wash contaminated clothing before reuse.
  • P321-Specific treatment (see on this label).
  • P501-Dispose of contents/container to accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.